missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB5 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77689AF594
This item is not returnable.
View return policy
Description
ABCB5 Polyclonal specifically detects ABCB5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ABCB5 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
| Rabbit | |
| Affinity Purified | |
| ABC Transporters, Lipid and Metabolism | |
| 340273.0 | |
| Store at 4°C in the dark. |
| Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ABCB5 | |
| A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 450-500) [UniProt Q2M3G0] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction