missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCB4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578691
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell.
The membrane-associated protein encoded ABCB4 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This gene encodes a full transporter and member of the p-glycoprotein family of membrane proteins with phosphatidylcholine as its substrate. The function of this protein has not yet been determined; however, it may involve transport of phospholipids from liver hepatocytes into bile. Alternative splicing of this gene results in several products of undetermined function.
Specifications
| ABCB4 | |
| Polyclonal | |
| Unconjugated | |
| ABCB4 | |
| ABC20; ABC21; ABCB4; ATP binding cassette subfamily B member 4; ATP-binding cassette sub-family B (MDR/TAP) member 4 (P-glycoprotein 3/ multidrug resistance 2; ATP-binding cassette sub-family B (MDR/TAP) member 4 (P-glycoprotein 3/ multidrug resistance 2); ATP-binding cassette sub-family B member 4; ATP-binding cassette, subfamily B (MDR/TAP), member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, sub-family B (MDR/TAP), member 4 (P-glycoprotein 3/ multidrug resistance 2); ATP-binding cassette, subfamily B, member 4; CD243; CLCS; GBD1; GP170; ICP3; MDR1; MDR2; mdr-2; MDR2/3; MDR3; MGC163296; Multidrug resistance protein 2; multidrug resistance protein 3; multiple drug resistance 3; P glycoprotein 2; P glycoprotein 3/multiple drug resistance 3; PFIC-3; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein 3/ multidrug resistance 2; P-glycoprotein-3/multiple drug resistance-3; P-GP; Pgp3; PGY1; Pgy2; Pgy-2; Pgy3; Phosphatidylcholine translocator ABCB4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5244 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P21439 | |
| ABCB4 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human ABCB4 (1-20aa MDLEAAKNGTAWRPTSAEGD). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction