missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCB10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530468
This item is not returnable.
View return policy
Description
Recommended positive controls: Raji, BCL-1. Predicted reactivity: Mouse (87%), Rat (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.
Specifications
| ABCB10 | |
| Polyclonal | |
| Unconjugated | |
| ABCB10 | |
| ABC transporter 10 protein; ABCB10; Abcb12; Abc-me; ABC-me protein; ABC-mitochondrial erythroid protein; ATP binding cassette subfamily B member 10; ATP-binding cassette sub-family B member 10, mitochondrial; ATP-binding cassette transporter 10; ATP-binding cassette, subfamily B (MDR/TAP), member 10; ATP-binding cassette, sub-family B (MDR/TAP), member 10; EST20237; M-ABC2; mitochondrial ATP-binding cassette 2; MTABC2; si:dkey-162b3.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23456, 56199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9JI39, Q9NRK6 | |
| ABCB10 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human ABCB10. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction