missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCB10 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563080

Product Code. 15730862

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MQTSGQRIVN RLRTSLFSSI LRQEVAFFDK TRTGELINRL SSDTALLGRS VTE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABCB10
Polyclonal
Unconjugated
ABCB10
ABC transporter 10 protein; ABCB10; Abcb12; Abc-me; ABC-me protein; ABC-mitochondrial erythroid protein; ATP binding cassette subfamily B member 10; ATP-binding cassette sub-family B member 10, mitochondrial; ATP-binding cassette transporter 10; ATP-binding cassette, subfamily B (MDR/TAP), member 10; ATP-binding cassette, sub-family B (MDR/TAP), member 10; EST20237; M-ABC2; mitochondrial ATP-binding cassette 2; MTABC2; si:dkey-162b3.3
Rabbit
Antigen Affinity Chromatography
RUO
23456
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NRK6
ABCB10
Recombinant protein corresponding to Human ABCB10. Recombinant protein control fragment (Product #RP-103209).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.