missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69065
This item is not returnable.
View return policy
Description
ABCB10 Polyclonal specifically detects ABCB10 in Rat samples. It is validated for Western Blot.
Specifications
| ABCB10 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ABC transporter 10 protein, ATP-binding cassette, sub-family B (MDR/TAP), member 10, EC 3.6.3, EC 3.6.3.44, EST20237, M-ABC2ATP-binding cassette transporter 10, Mitochondrial ATP-binding cassette 2, MTABC2ATP-binding cassette sub-family B member 10, mitochondrial | |
| Rabbit | |
| 77 kDa | |
| 100 μL | |
| Primary | |
| Zebrafish: 77%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q5FVL8 | |
| Abcb10 | |
| The immunogen for anti-Abcb10 antibody: synthetic peptide corresponding to a region of Rat (NP_001012166). Peptide sequence TRTGELINRLSSDTALLGHSVTENLSDGLRAVAQASVGVGMMFFVSPSLA. | |
| Affinity purified | |
| RUO | |
| 23456 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction