missing translation for 'onlineSavingsMsg'
Learn More

Abcam Sheep F(ab) Anti-Guinea pig IgG H&L (HRP), 500 uL

Product Code. 32109866 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109866 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109866 Supplier Abcam Supplier No. ab136791500uL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep F(ab) Anti-Guinea pig IgG H&L (HRP)

  • Host Species: Sheep
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, ICC/IF, IHC-Fr, IHC-P, WB
  • Species Reactivity: Guinea pig
  • Storage Buffer: Constituents: 50% Glycerol (glycerin, glycerine), 49% Tris buffered saline
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Constituents: 50% Glycerol (glycerin, glycerine), 49% Tris buffered saline
Host Species Sheep
Purification Method Affinity purification Immunogen
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Guinea pig.
Target Species Guinea Pig
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Sh
Shipping Condition Blue Ice
Reactivity abrv GP
Validated Application abrv ELISA, ICC/IF, IHC-Fr, IHC-P, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.