missing translation for 'onlineSavingsMsg'
Learn More

Abcam Sheep Anti-Mouse IgG H&L, 1 mg

Product Code. 32109961 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109961 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109961 Supplier Abcam Supplier No. ab67101mg
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Anti-Mouse IgG H&L

  • Host Species: Sheep
  • Clonality: Polyclonal
  • Applications: Conjugation, ELISA, ICC/IF, ID, IHC-Fr, IHC-P, IP, WB
  • Species Reactivity: Mouse
  • Storage Buffer: Potassium phosphate, Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Gene Symbols Ighg1
Host Species Sheep
Purification Method Affinity purification
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Liquid
Host abrv Sh
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv Conjugation, IHC-Fr, IHC-P, IP, ID, ICC/IF, WB, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.