missing translation for 'onlineSavingsMsg'
Learn More

Abcam Sheep Anti-Cow IgG heavy chain (HRP), 1 mg

Product Code. 32119393 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119393 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119393 Supplier Abcam Supplier No. ab1126181mg
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Anti-Cow IgG heavy chain (HRP)

  • Host Species: Sheep
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, ICC/IF, IHC-P, WB
  • Species Reactivity: Bovine
  • Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative,
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate HRP
Formulation pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative
Host Species Sheep
Purification Method Affinity purification Immunogen
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Cow.
Target Species Bovine
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Sh
Shipping Condition Blue Ice
Reactivity abrv Cow
Validated Application abrv ELISA, ICC/IF, IHC-P, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.