missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [SB77e] Anti-Mouse IgG1 Fc preadsorbed, 250 ug

Product Code. 32110713 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110713 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110713 Supplier Abcam Supplier No. ab99601250ug
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal antibody, suitable for Flow Cyt, ELISA.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: Flow Cyt, ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: pH: 8.2, Constituents: 100% Borate buffered saline
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone SB77e
Conjugate Unconjugated
Formulation pH: 8.2, Constituents: 100% Borate buffered saline
Gene Ighg1
Gene Symbols Ighg1
Host Species Rat
Purification Method Affinity purification Protein A
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Ighg1, Mouse
Validated Application abrv Flow Cyt, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.