missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [SB76b] Anti-Mouse IgG3 gamma chain (Alexa Fluor™ 647), 250 ug

Product Code. 32110971 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110971 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110971 Supplier Abcam Supplier No. ab172329250ug
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal antibody, suitable for ELISA, IHC-P, ICC/IF, Flow Cyt.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Applications: ELISA, IHC-P, ICC/IF, Flow Cyt
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ig gamma-3 chain C region
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SB76b
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Gene Ig gamma-3 chain C region
Host Species Rat
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG3
Host abrv Rt
Shipping Condition Blue Ice
Validated Application abrv ELISA, IHC-P, ICC/IF, Flow Cyt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.