missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [LO-MM-9] Anti-Mouse IgM heavy chain (Biotin), 250 ug

Product Code. 32117535 Shop All Abcam Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 μg
missing translation for 'unitSize':
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
32117535 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 32117535 missing translation for 'mfr' Abcam missing translation for 'supplierNo' ab11659250ug
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal antibody, suitable for ELISA.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG2a
  • Applications: ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: pH: 7.2, Preservative: 0.1% Sodium azide, Constituents: PBS
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA
Classification Monoclonal
Clone LO-MM-9
Conjugate Unconjugated
Formulation pH: 7.2, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene Ighm
Gene Symbols Ighm
Host Species Rat
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Packaging Vial
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2a
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Ighm, Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.