missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Rat monoclonal [LO-MG1-2] Anti-Mouse IgG1 heavy chain (Biotin), 250 ug
Shop All Abcam ProductsDescription
- Host Species: Rat
- Clonality: Monoclonal
- Isotype: IgG1
- Applications: ELISA
- Species Reactivity: Mouse
- Storage Buffer: pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
- Form: Liquid
- Purification: Affinity purification Protein G
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MG1-2 |
| Formulation | pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS |
| Gene | Ighg1 |
| Gene Symbols | Ighg1 |
| Host Species | Rat |
| Purification Method | Affinity purification Protein G |
| Quantity | 250 μg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?