missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rabbit monoclonal [M87-3] Anti-Mouse IgG1, IgG2a, IgG2b H&L - BSA and Azide free, 100 ug

Product Code. 32111345 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32111345 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32111345 Supplier Abcam Supplier No. ab288279100ug
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal antibody, suitable for ELISA.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: ELISA
  • Species Reactivity: Rat and Mouse
  • Storage Buffer: pH: 7.2 - 7.4, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA
Classification Monoclonal
Clone M87-3
Formulation pH: 7.2 - 7.4, Constituents: PBS
Gene Ighg1
Gene Symbols Ighg1
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rat and Mouse.
Target Species Rat, Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Rt, Ms
Target Molecule Ighg1, Rat, Mouse
Validated Application abrv ELISA
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.