missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Host Species: Rabbit
- Clonality: Polyclonal
- Isotype: IgG
- Applications: IHC-P, ICC, ELISA, WB
- Species Reactivity: Sheep
- Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative,
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative |
| Host Species | Rabbit |
| Purification Method | Affinity purification Immunogen |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?