missing translation for 'onlineSavingsMsg'
Learn More

Abcam Native Cow Osteocalcin protein, 100 ug

Product Code. 32144104 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32144104 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32144104 Supplier Abcam Supplier No. ab62262100ug
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for RIA, ELISA, Dot, SDS-PAGE.

  • Applications: RIA, ELISA, Dot, SDS-PAGE
  • Storage Buffer: pH: 7.4, Constituents: 50% Glycerol (glycerin, glycerine), 0.435% Sodium chloride, 0.121% Tris
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Formulation pH: 7.4, Constituents: 50% Glycerol (glycerin, glycerine), 0.435% Sodium chloride, 0.121% Tris
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.