missing translation for 'onlineSavingsMsg'
Learn More

Abcam Native Cow Bovine Serum Albumin protein, 50 g

Product Code. 32173347 Shop All Abcam Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32173347 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32173347 Supplier Abcam Supplier No. ab6400950g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for WB, IHC, with 8 citations.

  • Applications: WB, IHC
  • Form: Lyophilized

Specifications

Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Gene Symbols ALB
Quantity 50 g
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Target Species Non-species specific
Product Type Secondary Antibody
Form Lyophilized
Shipping Condition Ambient
Validated Application abrv WB, IHC
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.