missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Mouse Von Willebrand Factor A2 Matched Antibody Pair Kit, 5 x 96 Tests
Shop All Abcam ProductsDescription
- Applications: ELISA, IA
- Species Reactivity: Mouse
Specifications
Specifications
| Applications | ELISA |
| Conjugate | Unconjugated |
| Gene Symbols | Trmt10a |
| Quantity | 5 x 96 Tests |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Test Specificity | This product reacts with Mouse. |
| Target Species | Mouse |
| Product Type | Secondary Antibody |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?