missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mouse/Rat MMP13 Antibody Pair - BSA and Azide free, 10 x 96 Tests

Product Code. 32115450 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115450 10 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115450 Supplier Abcam Supplier No. ab31445810x96Tests
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for sELISA.

  • Applications: sELISA
  • Species Reactivity: Mouse and Rat

Tekniske data

Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols Mmp13
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse and Rat.
Target Species Mouse, Rat
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt
Validated Application abrv sELISA
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.