missing translation for 'onlineSavingsMsg'
Learn More

Abcam Human/Mouse TRAP/CD40L Antibody Pair - BSA and Azide free (TNFSF5), 10 x 96 Tests

Product Code. 32119897 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119897 10 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32119897 Supplier Abcam Supplier No. ab25343610x96Tests
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for sELISA.

  • Applications: sELISA
  • Species Reactivity: Mouse and Human

Specifications

Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols Cd40lg
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse and Human.
Target Species Mouse, Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Ms, Hu
Validated Application abrv sELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.