missing translation for 'onlineSavingsMsg'
Learn More

Abcam Bovine Heart Mitochondria, 2 mg

Product Code. 32156732 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32156732 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32156732 Supplier Abcam Supplier No. ab1103382mg
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for WB, SDS-PAGE, IP, with 3 citations.

  • Applications: WB, SDS-PAGE, IP
  • Storage Buffer: Constituents: 99.98% Isolation Buffer, 0.02% PMSF
  • Form: Liquid

Specifications

Applications Western Blot, SDS-Page, Immunoprecipitation
Conjugate Unconjugated
Formulation Constituents: 99.98% Isolation Buffer, 0.02% PMSF
Quantity 2 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Target Species Non-species specific
Product Type Secondary Antibody
Form Liquid
Shipping Condition Blue Ice
Validated Application abrv WB, SDS-PAGE, IP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.