missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-mouse IgG for IP (HRP), 100 uL

Product Code. 32117575 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117575 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117575 Supplier Abcam Supplier No. ab131368100uL
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal secondary antibody, suitable for WB, IP, ELISA, with over 100 citations.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: WB, IP, ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: Constituents: 50% Glycerol (glycerin, glycerine), 0.88% Sodium chloride, 0.16% Sodium phosphate, 0.1% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein G

Specifications

Applications Western Blot, Immunoprecipitation, ELISA
Classification Monoclonal
Clone eB144
Conjugate HRP
Formulation Constituents: 50% Glycerol (glycerin, glycerine), 0.88% Sodium chloride, 0.16% Sodium phosphate, 0.1% BSA
Gene Symbols Ighg1
Host Species Rat
Purification Method Affinity purification Protein G
Quantity 100 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv WB, IP, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.