missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-mouse IgG for IP (Biotin), 50 ug

Product Code. 32115227 Shop All Abcam Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32115227 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32115227 Supplier Abcam Supplier No. ab13136750ug
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal secondary antibody, suitable for WB, with 2 citations.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: WB
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: 0.88% Sodium chloride, 0.16% Sodium phosphate
  • Form: Liquid

Specifications

Applications Western Blot
Classification Monoclonal
Clone eB144
Conjugate Biotin
Formulation Preservative: 0.09% Sodium azide, Constituents: 0.88% Sodium chloride, 0.16% Sodium phosphate
Gene Symbols Ighg1
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.