missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-Collagen IV antibody, 100 ug

Product Code. 32106618 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32106618 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32106618 Supplier Abcam Supplier No. ab6586100ug
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal antibody, suitable for IHC-P, IP, ELISA, WB, with over 1,000 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: IHC-P, IP, ELISA, WB, IF, IHC-FoFr, ICC/IF, IHC-Fr, IHC-FrFl
  • Species Reactivity: Human, Hamster, Syrian hamster, Cow, Mouse, Dog, Zebrafish, Rat, Chinese hamster, African green monkey and Pig
  • Storage Buffer: pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
  • Form: Liquid
  • Purification: Affinity purification Immunogen

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen COL4A1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, ELISA, Western Blot, Immunofluorescence, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry (Free Floating)
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Gene COL4A1
Gene Symbols COL4A1
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 100 μg
Regulatory Status RUO
Chains or Fragments IgG
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human, Hamster, Syrian hamster, Cow, Mouse, Dog, Zebrafish, Rat, Chinese hamster, African green monkey and Pig.
Target Species Human, Hamster, Syrian Hamster, Bovine, Mouse, Dog, Zebrafish, Rat, Chinese Hamster, African Green Monkey, Pig
Product Type Polyclonal Antibody (COL4A1)
Form Liquid
Format Vial
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Hu, Ha, Cow, Ms, Dog, Zf, Rt, CHa, AGMk, Pig
Target Gene COL4A1
Target Entry COL4A1
Target Molecule COL4A1, Human, Hamster, Syrian hamster, Cow, Mouse, Dog, Zebrafish, Rat, Chinese hamster, African green monkey, Pig
Technique IHC-P, IP, ELISA, WB, IF, IHC-FoFr, ICC/IF, IHC-Fr, IHC-FrFl
Validated Application abrv IHC-P, IP, ELISA, WB, IF, IHC-FoFr, ICC/IF, IHC-Fr, IHC-FrFl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.