missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-CD31 antibody [JC/70A], 100 ug

Product Code. 32047302 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32047302 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32047302 Supplier Abcam Supplier No. ab9498100ug
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal antibody, suitable for IHC-P, Flow Cyt, WB, IHC-Fr, with over 400 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: IHC-P, Flow Cyt, WB, IHC-Fr, ICC/IF
  • Species Reactivity: Human
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 6.97% L-Arginine
  • Form: Liquid
  • Purification: Affinity purification

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen PECAM1
Applications Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone JC/70A
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 6.97% L-Arginine
Gene PECAM1
Gene Symbols PECAM1
Host Species Mouse
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Chains or Fragments IgG1
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Antibody (PECAM1)
Form Liquid
Format Vial
Isotype IgG1
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Gene PECAM1
Target Entry PECAM1
Target Molecule PECAM1, Human
Technique IHC-P, Flow Cyt, WB, IHC-Fr, ICC/IF
Validated Application abrv IHC-P, Flow Cyt, WB, IHC-Fr, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.