missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24351-1-AP-150UL
This item is not returnable.
View return policy
Description
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of proteins that catalyze the transport of molecules across extracellular and intracellular membranes by harnessing the energy of ATP hydrolysis. ABCA8 (ATP-binding cassette, sub-family A (ABC1), member 8) is a 1, 581 amino acid multi-pass membrane protein that contains two ABC transporter domains and belongs to the ABC transporter family. Expressed in a variety of tissues with particularly high expression in liver, heart and skeletal muscle, ABCA8 functions as an ATP-dependent lipophilic drug transporter that exists as multiple alternatively spliced isoforms.Specifications
| ABCA8 | |
| Polyclonal | |
| Unconjugated | |
| ABCA8 | |
| ABCA8, KIAA0822 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10351, 217258, 27404 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94911, Q8K440, Q8K442 | |
| ABCA8, Abca8a, Abca8b | |
| ABCA8 Fusion Protein Ag18991 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction