missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total Abca6.
The ATP-binding cassette (ABC) superfamily is comprised of transmembrane proteins involved in energy-dependent transport of a variety of substrates across membranes. ABCA6 is a 1,617 amino acid protein belonging to the ABC transporter family. ABCA6 is up-regulated during monocyte differentiation into macrophages, suggesting a role in macrophage lipid homeostasis. ABCA6 contains two ABC transporter domains and is expressed as three isoforms produced by alternative splicing. ABCA6 is present at highest levels in adult liver, with significant levels found in many adult tissues and fetal lung, kidney and liver.
Specifications
Specifications
| Antigen | ABCA6 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ABCA6 |
| Gene Accession No. | Q8K441, Q8N139 |
| Gene Alias | 6330565N06Rik; ABC transporter ABCA6; Abca6; ATP binding cassette subfamily A member 6; ATP-binding cassette A6; ATP-binding cassette sub-family A member 6; ATP-binding cassette transporter sub-family A member 6; ATP-binding cassette, subfamily A (ABC1), member 6; ATP-binding cassette, sub-family A (ABC1), member 6; EST155051 |
| Gene Symbols | ABCA6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
