missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578688
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, rat eye tissue, mouse eye tissue.
ABCA4 (ATP-Binding Cassette, Subfamily A, Member 4), also known as ABCR, is a protein which in humans is encoded by the ABCA4 gene. ABCA4 is a member of the ATP-binding cassette transporter gene sub-family A (ABC1) found exclusively in multicellular eukaryotes. Using a whole genome radiation hybrid panel, this gene is mapped to 1p21-p13. And this gene is expressed exclusively in retina photoreceptor cells, indicating the gene product mediates transport of an essental molecule across the photoreceptor cell membrane. Additionally, it is showed by immunofluorescence microscopy and Western blot analysis that ABCR is present in foveal and peripheral cone, as well as rod, photoreceptors. The results suggested that the loss in central vision experienced by patients with Stargardt macular dystrophy arises directly from ABCR-mediated foveal cone degeneration.
Specifications
| ABCA4 | |
| Polyclonal | |
| Unconjugated | |
| ABCA4 | |
| ABC transporter; ABC10; abca4; abca4.L; Abcr; ARMD2; ATP binding cassette subfamily A member 4; ATP binding cassette subfamily A member 4 L homeolog; ATP binding cassette transporter; ATP-binding cassette 10; ATP-binding cassette sub-family A member 4; ATP-binding cassette transporter, retinal-specific; ATP-binding cassette, subfamily A (ABC1), member 4; ATP-binding cassette, sub-family A (ABC1), member 4; ATP-binding cassette, sub-family A, member 4; ATP-binding transporter, retina-specific; AW050280; CORD3; D430003I15Rik; FFM; photoreceptor rim protein; retinal ABCA4 transporter; retinal-specific ATP-binding cassette transporter; Retinal-specific phospholipid-transporting ATPase ABCA4; retina-specific ABC transporter; RIM ABC transporter; RIM protein; RMP; RP19; Stargardt disease protein; STGD; STGD1; XELAEV_180233141mg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11304, 24, 310836 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| O35600, P78363 | |
| ABCA4 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human ABCA4 (1890-1927aa FLLTLLVQRHFFLSQWIAEPTKEPIVDEDDDVAEERQR). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction