missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560157
This item is not returnable.
View return policy
Description
Immunogen sequence: PTISVKEAFY TAAPLTSAGI LPVMQSLCPD GQRDEFGFLQ YANSTVTQLL ERLDRVVEEG NLFDPARPSL GSELEALRQH LEALSAGPGT SGSHLDRSTV SSFSLDSV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.
Specifications
| ABCA2 | |
| Polyclonal | |
| Unconjugated | |
| Abca2 | |
| Abc2; ABCA2; abca2 {ECO:0000312; AI413825; ATP binding cassette subfamily A member 2; ATP-binding cassette 2; ATP-binding cassette sub-family A member 2; ATP-binding cassette transporter 2; ATP-binding cassette, subfamily A (ABC1), member 2; ATP-binding cassette, sub-family A (ABC1), member 2; ATP-binding cassette, sub-family A, member 2; D2H0S1474E; KIAA1062; mKIAA1062; RGD:620238} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZC7 | |
| Abca2 | |
| Recombinant protein corresponding to Human ABCA2. Recombinant protein control fragment (Product #RP-92911). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction