missing translation for 'onlineSavingsMsg'
Learn More

ABCA13 Antibody (2379B) [Biotin], Novus Biologicals™

Product Code. 30664425 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30664425 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30664425 Supplier Novus Biologicals Supplier No. FAB11195B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ABCA13 Monoclonal antibody specifically detects ABCA13 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA13
Applications Immunohistochemistry
Classification Monoclonal
Clone 2379B
Conjugate Biotin
Formulation PBS
Gene Alias ATP binding cassette transporter A13, ATP-binding cassette sub-family A member 13, ATP-binding cassette, sub-family A (ABC1), member 13, DKFZp313D2411, EC 2.7.11.25, EC 3.6.3, FLJ16398, FLJ33876, FLJ33951
Host Species Rabbit
Immunogen Synthetic peptide within aminoacids 4700-4750 of the Human ABCA13 protein,, Accession # Q86UQ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154664
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.