missing translation for 'onlineSavingsMsg'
Learn More

ABCA10 Monoclonal Antibody (8F4)
GREENER_CHOICE

Product Code. 15588942
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15588942 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588942 Supplier Invitrogen Supplier No. MA521013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ABCA10 Monoclonal Antibody (8F4)

ABCA10 Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA10
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ABCA10
Gene Accession No. Q8WWZ4
Gene Alias ABCA10; ATP binding cassette subfamily A member 10; ATP-binding cassette A10; ATP-binding cassette sub-family A member 10; ATP-binding cassette, sub-family A (ABC1), member 10; EST698739; FKHL6; FREAC2
Gene Symbols ABCA10
Host Species Mouse
Immunogen ABCA10 (NP_525021, 1 a.a. ~ 80 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.