missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98086-50ul
This item is not returnable.
View return policy
Description
ABC2 Polyclonal antibody specifically detects ABC2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ABC2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| ABC2, Amplified in breast cancer gene 2 protein, chromosome 17 open reading frame 71, FLJ10587, protein SMG8, Protein smg-8 homolog, SMG8 | |
| Produced in rabbits immunized with E. coli-derived Human ABC2 fragment. | |
| 50 μg | |
| Cancer | |
| 55181 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction