missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111139
This item is not returnable.
View return policy
Description
Immunogen sequence: LSVAPKGMSQ LITMACGSCS NENALKTIFM WYRSKERGQR GFSQEELETC MINQAPGCPD YSILSFMGAF HGRTMGCLAT THSKAIHKI Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| ABAT | |
| Polyclonal | |
| Unconjugated | |
| ABAT | |
| (S)-3-amino-2-methylpropionate transaminase; 4-aminobutyrate aminotransferase; 4-aminobutyrate aminotransferase, brain isoform; 4-aminobutyrate aminotransferase, liver isoform; 4-aminobutyrate aminotransferase, mitochondrial; 4-aminobutyrate transaminase; 9630038C02Rik; ABAT; AI255750; beta-AlaAT; beta-alanine oxoglutarate aminotransferase; cb880; ENSMUSG00000051226; fj82a01; FLJ17813; FLJ30272; GABA aminotransferase; GABA AT; GABA T; GABA transaminase; GABA transferase; Gabaat; GABA-AT; Gabat; GABA-T; Gamma-amino-N-butyrate transaminase; Gm9851; hCG1984265; I54; L AIBAT; Laibat; L-AIBAT; mitochondrial 4-aminobutyrate aminotransferase; NPD009; wu:fj82a01; X61497 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P80404 | |
| ABAT | |
| Recombinant Protein corresonding to Human ABAT. Recombinant protein control fragment (Product #RP-98784). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction