missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABAT Rabbit anti-Human, Mouse, Rat, Clone: 10V1K9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | ABAT |
|---|---|
| Clone | 10V1K9 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384914
|
Novus Biologicals
NBP3-16767-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18339314
|
Novus Biologicals
NBP3-16767-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABAT Monoclonal antibody specifically detects ABAT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ABAT | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| (S)-3-amino-2-methylpropionate transaminase, 4-aminobutyrate aminotransferase, 4-aminobutyrate aminotransferase, mitochondrial, 4-aminobutyrate transaminase, EC 2.6.1.19, EC 2.6.1.22, FLJ17813, FLJ30272, G, GABA aminotransferase, GABA transaminase, GABA transferase, GABA-AT, GABA-T, GABATNPD009, Gamma-amino-N-butyrate transaminase, hCG1984265, L-AIBAT | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ABAT (P80404). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 10V1K9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 18 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title