missing translation for 'onlineSavingsMsg'
Learn More

Abarelix Trifluoroacetate Salt (~90%), TRC

Product Code. 30317843
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317843 1 mg 1mg
30312114 5 mg 5mg
30315399 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30317843 Supplier Toronto Research Chemicals Supplier No. TRCA1072251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abarelix Trifluoroacetate Salt
Synonym N-Acetyl-3-(2-naphthalenyl)-D-alanyl-4-chloro-D-phenylalanyl-3-(3-pyridinyl)-D-alanyl-L-seryl-N-methyl-L-tyrosyl-D-asparaginyl-L-leucyl-N6-(1-methylethyl)-L-lysyl-L-prolyl-D-alaninamide trifluoroacetate salt∣17: PN: WO2007012430 SEQID: 19 claimed protein trifluoroacetate salt∣1: PN: WO02056903 PAGE: 25 claimed protein trifluoroacetate salt∣5: PN: WO0018423 PAGE: 26 claimed protein trifluoroacetate salt∣Abarelix trifluoroacetate salt∣PPI 149 trifluoroacetate salt∣Plenaxis trifluoroacetate salt∣R 3827 trifluoroacetate salt∣Abarelix TFA
Quantity 1 mg
Shipping Condition Room Temperature
Molecular Formula C72H95ClN14O14 . x(C2HF3O2)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.