missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Abarelix

Product Code. 16473366
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473366 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16473366 Supplier MedChemExpress Supplier No. HY13534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Abarelix (R3827; PPI 149) is a potent gonadotrophin-releasing hormone (GnRH) antagonist, used for prostate cancer treatment.

Specifications

Purity 96.27%
CAS 183552-38-7
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 1416.06 g/mol
Product Type Peptides
Quantity 25 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 14.2 mg/mL (10.03 mM)
Synonym R3827∣ PPI 149
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.