missing translation for 'onlineSavingsMsg'
Learn More

Abamectin

Product Code. 15482879
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15482879

Brand: Thermo Scientific Alfa Aesar J60039.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interacts with GABA receptors to cause chloride channel activation | CAS: 71751-41-2 | C48H72O14

Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

Solubility
Soluble in DMSO

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71751-41-2
Molecular Formula C48H72O14
MDL Number MFCD01769550
Synonym Avermectin B1

Specifications

Melting Point 150°C to 155°C
Color White to Yellow
Quantity 5 g
UN Number UN2811
Merck Index 14,2
Solubility Information Soluble in DMSO
Formula Weight 873.1
Physical Form Powder
Chemical Name or Material Abamectin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.