missing translation for 'onlineSavingsMsg'
Learn More

Abacavir Sulfate EP Impurity C (Abacavir Related USP Compound A), Mikromol™

Product Code. 30052515
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052515 25 mg 25mg
30051132 100 mg 100mg
30461700 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052515 Supplier Mikromol Supplier No. MM3249.010025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Abacavir Sulfate EP Impurity C (Abacavir Related USP Compound A)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Descyclopropyl Abacavir
CAS 124752-25-6
Analyte or Component Names Descyclopropyl Abacavir
Shipping Condition Room Temperature
Molecular Formula C11 H14 N6 O
Certifications/Compliance ISO 17025
Synonym [(1S,4R)-4-(2,6-Diamino-9H-purin-9-yl)cyclopent-2-enyl]methanol, 2-Cyclopentene-1-methanol, 4-(2,6-diamino-9H-purin-9-yl)-, (1S,4R)-, 2-Cyclopentene-1-methanol, 4-(2,6-diamino-9H-purin-9-yl)-, (1S-cis)-, Descyclopropyl Abacavir, Abacavir Related Compound A (USP), Abacavir Sulfate Imp. C (Ph. Int.), Abacavir Sulfate Imp C (EP), Abacavir Imp. C (EP), Abacavir USP Related Compound A, Abacavir USP RC A, 139U91 (GR92685X), GW 466608X, GW 470921X, GW 403126X, GW 357662X, GW 444710X, Abacavir Related Compound A, Abacavir Sulfate Impurity C, Abacavir Impurity C
InChI Formula InChI=1S/C11H14N6O/c12-9-8-10(16-11(13)15-9)17(5-14-8)7-2-1-6(3-7)4-18/h1-2,5-7,18H,3-4H2,(H4,12,13,15,16)/t6-,7+/m1/s1
SMILES Nc1nc(N)c2ncn([C@@H]3C[C@H](CO)C=C3)c2n1
IUPAC Name [(1S,4R)-4-(2,6-diaminopurin-9-yl)cyclopent-2-en-1-yl]methanol
Molecular Weight (g/mol) 246.27
Quantity 25 mg
Formula Weight 246.1229 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.