missing translation for 'onlineSavingsMsg'
Learn More

Abacavir sulfate, 99 %

Product Code. 15519463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15519463 1 g 1g
15529463 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15519463 Supplier Thermo Scientific Acros Supplier No. 458860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 188062-50-2 | C14H18N61/2H2SO4 | 335.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188062-50-2
Molecular Formula C14H18N61/2H2SO4
Molecular Weight (g/mol) 335.37
InChI Key WMHSRBZIJNQHKT-NHMRBCHTSA-N
Synonym abacavir hemisulfate, 1s,4r-4-2-amino-6-cyclopropylamino-9h-purin-9-yl-2-cyclopentene-1-methanol hemisulfate
PubChem CID 75539558
IUPAC Name [(1S)-4-[2-amino-6-(cyclopropylamino)purin-9-yl]cyclopent-2-en-1-yl]methanol;sulfuric acid
SMILES C1CC1NC2=NC(=NC3=C2N=CN3C4CC(C=C4)CO)N.C1CC1NC2=NC(=NC3=C2N=CN3C4CC(C=C4)CO)N.OS(=O)(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 335.37
Percent Purity 99%
Chemical Name or Material Abacavir sulfate
compliance-icons compliance-icons
Product Identifier
  • Abacavir sulfate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.