missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Abacavir hemisulfate

Product Code. 18730268
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18730268 10 mg 10mg
18766818 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 18730268

Brand: Tocris Bioscience™ 4148/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reverse transcriptase inhibitor; antiretroviral

Abacavir hemisulfate is a competitive reverse transcriptase inhibitor; inhibits HIV replication. Orally active and brain penetrant. Antiretroviral agent.
TRUSTED_SUSTAINABILITY

Specifications

Purity 99%
Molecular Formula (1 S,4 R)-4-[2-Amino-6-(cyclopropylamino)-9 H-purin-9-yl]-2-cyclopentene-1-methanol hemisulfate
CAS 188062-50-2
Content And Storage 4°C
Quantity 10 mg
Product Type Abacavir hemisulfate
Molecular Weight (g/mol) M.W. (Observed): 335.37
Inhibitors Abacavir hemisulfate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.