missing translation for 'onlineSavingsMsg'
Learn More

Abacavir Carboxylate, TRC

Product Code. 30179388
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179388 10 mg 10mg
30179424 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179388 Supplier Toronto Research Chemicals Supplier No. TRCA10500510MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 384380-52-3
Molecular Formula C14 H16 N6 O2
Synonym 2-Cyclopentene-1-carboxylic acid, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1S,4R)-∣(1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-carboxylic acid∣2269W∣Abacavir Carboxylate

Specifications

Format Neat
Molecular Weight (g/mol) 300.32
Shipping Condition Room Temperature
Quantity 10 mg
Chemical Name or Material Abacavir Carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.