missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAV4 VP3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93576JF549
This item is not returnable.
View return policy
Description
AAV4 VP3 Polyclonal antibody specifically detects AAV4 VP3 in Nonspecies specific, Virus samples. It is validated for Western Blot
Specifications
| AAV4 VP3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| Adenovirus 4, AdV 4 | |
| Directed against a synthetic peptide within residues 450-475 of 81 kDa capsid VP3 protein of AAV 4. | |
| 0.1 mL | |
| Gene Therapy, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| All species, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction