missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AATK/Serine/threonine-protein kinase LMTK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | AATK/Serine/threonine-protein kinase LMTK1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453982
|
Novus Biologicals
NBP1-90292-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205636
|
Novus Biologicals
NBP1-90292 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AATK/Serine/threonine-protein kinase LMTK1 Polyclonal specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AATK/Serine/threonine-protein kinase LMTK1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1EC 2.7.11.1, LMTK1AATYK1, serine/threonine-protein kinase LMTK1 | |
| AATK | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9625 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TSGIFTDTSSDGLQARRPDVVPAFRSLQKQVGTPDSLDSLDIPSSASDGGYEVFSPSATGPSGGQPRALDSGYDTENYESPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title