missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AATK/Serine/threonine-protein kinase LMTK1 Antibody (456006), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4866X
This item is not returnable.
View return policy
Description
AATK/Serine/threonine-protein kinase LMTK1 Monoclonal antibody specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| AATK/Serine/threonine-protein kinase LMTK1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 456006 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1EC 2.7.11.1, LMTK1AATYK1, serine/threonine-protein kinase LMTK1 | |
| E. coli-derived recombinant human AATK, Asn421-Ala589, Accession # AAH47378 | |
| 0.1 mL | |
| Protein Kinase | |
| 9625 | |
| Store at 4°C in the dark. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido