missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AATK/Serine/threonine-protein kinase LMTK1 Antibody (2C8), Novus Biologicals™

Product Code. 18347858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347858 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347858 Supplier Novus Biologicals™ Supplier No. H00009625M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AATK/Serine/threonine-protein kinase LMTK1 Monoclonal antibody specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AATK/Serine/threonine-protein kinase LMTK1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH47378
Gene Alias AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1EC 2.7.11.1, LMTK1AATYK1, serine/threonine-protein kinase LMTK1
Host Species Mouse
Immunogen AATK (AAH47378, 161 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSAYFSDLEAEAEATSGPEKKCGGDRAPGPELGLRSTGQPSEQVCLRPGVSG
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9625
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.