missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AATF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76655
This item is not returnable.
View return policy
Description
AATF Polyclonal specifically detects AATF in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AATF | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| apoptosis antagonizing transcription factor, Apoptosis-antagonizing transcription factor, CHE-1, CHE1Rb-binding protein Che-1, DEDprotein AATF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| NP_036270 | |
| AATF | |
| Antibody was raised against a 12 amino acid synthetic peptide from near the carboxy terminus of human AATF. The immunogen is located within the last 50 amino acids of AATF. Amino Acid Squence: GQLHPPDEGHGD | |
| 0.1 mg | |
| Apoptosis | |
| 26574 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction