missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AASS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545462
This item is not returnable.
View return policy
Description
Peptide sequence: HHHLVRKTDA VYDPAEYDKH PERYISRFNT DIAPYTTCLI NGIYWEQNTP Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rabbit: 86%; Rat: 86%.
AASS is a bifunctional enzyme that catalyzes the first two steps in the mammalian lysine degradation pathway. The N-terminal and the C-terminal portions of this enzyme contain lysine-ketoglutarate reductase and saccharopine dehydrogenase activity, respectively, resulting in the conversion of lysine to alpha-aminoadipic semialdehyde. Mutations in the gene encoding this protein are associated with familial hyperlysinemia.
Specifications
| AASS | |
| Polyclonal | |
| Unconjugated | |
| AASS | |
| AASS; aass {ECO:0000312; alpha-aminoadipate semialdehyde synthase; alpha-aminoadipic semialdehyde synthase, mitochondrial; aminoadipate-semialdehyde synthase; aminoadipic semialdehyde synthase; EMBL:AAI28772.1}; LKR; LKR/SDH; LKRSDH; LOR; LOR/SDH; Lorsdh; Lysine ketoglutarate reductase; lysine oxoglutarate reductase; lysine-2-oxoglutarate reductase; lysine-ketoglutarate reductase /saccharopine dehydrogenase; saccharopine dehydrogenase; SDH | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10157 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UDR5 | |
| AASS | |
| Synthetic peptide directed towards the C-terminal of human AASS (aa 268-317). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction