missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AASS Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92093-0.1ml
This item is not returnable.
View return policy
Description
AASS Polyclonal antibody specifically detects AASS in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| AASS | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| aminoadipate-semialdehyde synthase, aminoadipic semialdehyde synthase, LKR/SDH, LKRSDH, LORSDH, lysine-2-oxoglutarate reductase, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 180-400 of human AASS (NP_005754.2). MHIGMAHNYRNSSQAVQAVRDAGYEISLGLMPKSIGPLTFVFTGTGNVSKGAQAIFNELPCEYVEPHELKEVSQTGDLRKVYGTVLSRHHHLVRKTDAVYDPAEYDKHPERYISRFNTDIAPYTTCLINGIYWEQNTPRLLTRQDAQSLLAPGKFSPAGVEGCPALPHKLVAICDISADTGGSIEFMTECTTIEHPFCMYDADQHIIHDSVEGSGILMCSI | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 10157 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction