missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AASDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585227
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: Huh 7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AASDH encodes a member of the non-ribosome peptide syntesase (NRPS) enzyme family. The encoded protein contains an AMP-binding domain, PP-binding (phosphopantetheine, or pantetheine 4'phosphate-binding) domain and the Pyrrolo-quinoline quinon (PQQ) binding domain. The protein is expressed in several adult tissues. Covalently binds beta-alanine in an ATP-dependent manner to form a thioester bond with its phosphopantetheine group and transfers it to an, as yet, unknown acceptor. May be required for a post-translational protein modification or for post-transcriptional modification of an RNA.
Specifications
| AASDH | |
| Polyclonal | |
| Unconjugated | |
| AASDH | |
| 2-aminoadipic 6-semialdehyde dehydrogenase; 2-aminoadipic 6-semialdehyde dehydrogenase antibody; A230062G08Rik; A830035E16; Aasdh; AASDH antibody; ACSF4; ACSF4 antibody; acyl-CoA synthetase family member 4; acyl-CoA synthetase family member 4 antibody; aminoadipate-semialdehyde dehydrogenase; aminoadipate-semialdehyde dehydrogenase antibody; Beta-alanine-activating enzyme; HSPC318; LYS2; LYS2 antibody; non-ribosomal peptide synthetase 1098; non-ribosomal peptide synthetase 1098 antibody; non-ribosomal peptide synthetase 998; non-ribosomal peptide synthetase 998 antibody; NRPS1098; NRPS1098 antibody; NRPS998; NRPS998 antibody; Protein LYS2 homolog; Protein NRPS998; putative aminoadipate-semialdehyde dehydrogenase; RGD1311135; U26 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 132949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q4L235 | |
| AASDH | |
| Recombinant protein encompassing a sequence within the N-terminus region of human AASDH. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction