missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AASDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | AASDH |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453911
|
Novus Biologicals
NBP1-89266-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18275446
|
Novus Biologicals
NBP1-89266 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AASDH Polyclonal specifically detects AASDH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AASDH | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ACSF4acyl-CoA synthetase family member 4, aminoadipate-semialdehyde dehydrogenase, EC 6.2.1.-, LYS2, non-ribosomal peptide synthetase 1098, non-ribosomal peptide synthetase 998,2-aminoadipic 6-semialdehyde dehydrogenase, NRPS1098, NRPS998, Protein NRPS998, U26 | |
| AASDH | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q4L235 | |
| 132949 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RKLSDINQEEASGTSLHQKAIMTFTCHNEINAFVVLSRGSQILSLNSTRFLTKLGHCSSACPSDSVSQTNIQNLKGLNSPVLIGKSKDPSCV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title