missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AASD-PPT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £359.00
Specifications
| Antigen | AASD-PPT |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624962
|
Novus Biologicals
NBP2-92449-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611562
|
Novus Biologicals
NBP2-92449-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AASD-PPT Polyclonal antibody specifically detects AASD-PPT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AASD-PPT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Proteases & Other Enzymes | |
| PBS with 50% glycerol, pH7.3. | |
| 60496 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 4'-phosphopantetheinyl transferase, AASD-PPTDKFZp566E2346, Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase, aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, CGI-80, EC 2.7.8, EC 2.7.8.-, L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, LYS2, LYS5, LYS5 ortholog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-309 of human AASDHPPT (NP_056238.2). MVFPAKRFCLVPSMEGVRWAFSCGTWLPSRAEWLLAVRSIQPEEKERIGQFVFARDAKAAMAGRLMIRKLVAEKLNIPWNHIRLQRTAKGKPVLAKDSSNPYPNFNFNISHQGDYAVLAAEPELQVGIDIMKTSFPGRGSIPEFFHIMKRKFTNKEWETIRSFKDEWTQLDMFYRNWALKESFIKAIGVGLGFELQRLEFDLSPLNLDIGQVYKETRLFLDGEEEKEWAFEESKIDEHHFVAVALRKPDGSRHQDVPSQDDSKPTQRQFTILNFNDLMSSAVPMTPEDPSFWDCFCFTEEIPIRNGTKS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title